Order Entry
Export
ContactUsLinkComponent
Anti-TFRC Mouse Monoclonal Antibody [clone: MRQ-48]
Anti-TFRC Mouse Monoclonal Antibody [clone: MRQ-48]
Catalog # ORIGTA327652
Supplier:  OriGene
Anti-TFRC Mouse Monoclonal Antibody [clone: MRQ-48]
Catalog # ORIGTA327652
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TFRC
  • Clonality:
    Monoclonal
  • Clone:
    MRQ-48
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    The antibody is diluted in tris buffered saline, pH 7,3-7.7, with 1% BSA and <1% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Purification:
    purified by affinity chromatography
  • Pk:
    1 mL

Specifications

About this item

Recommended Dilutions: Immunochemistry: 1:100-1:500

Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: MRQ-48
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human