Order Entry
Export
ContactUsLinkComponent
Anti-MEK1 Rabbit Polyclonal Antibody
Anti-MEK1 Rabbit Polyclonal Antibody
Catalog # ORIGTA326799
Supplier:  OriGene
Anti-MEK1 Rabbit Polyclonal Antibody
Catalog # ORIGTA326799
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MEK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    Store at -20C or -80C. Avoid freeze / thaw cycles. Buffer: PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    C term -peptide of human MEK1
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat