Anti-FAM129A Rabbit Polyclonal Antibody
Catalog # ORIGTA327590
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:FAM129A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Modification:phospho-Ser602
- Storage buffer:Store at -20C or -80C. Avoid freeze / thaw cycles. Buffer: PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:A phospho specific peptide corresponding to residues surrounding S602 of human FAM129A
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: FAM129A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human