Order Entry
Export
ContactUsLinkComponent
Anti-PXN Rabbit Polyclonal Antibody
Anti-PXN Rabbit Polyclonal Antibody
Catalog # ORIGTA326090
Supplier:  OriGene
Anti-PXN Rabbit Polyclonal Antibody
Catalog # ORIGTA326090
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Paxillin
  • Antigen symbol:
    PXN
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    The antiserum was produced against synthesized non-phosphopeptide derived from human Paxillin around the phosphorylation site of serine 272 (M-A-SP-L-S).
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:20000

Type: Primary
Antigen: PXN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat