Anti-TUBA4A Rabbit Polyclonal Antibody
ORIGTA326047
: OriGene
- Antibody type:Primary
- Antigen symbol:TUBA4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Modification:phospho-Tyr272
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:The antiserum was produced against synthesized phosphopeptide derived from human TUBA1/3/4 around the phosphorylation site of tyrosine 272 (A-T-YpA-P).
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-1:3000 ELISA: 1:40000
Type: Primary
Antigen: TUBA4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat