Order Entry
Export
ContactUsLinkComponent
Anti-ATF2 Rabbit Polyclonal Antibody
Anti-ATF2 Rabbit Polyclonal Antibody
Catalog # ORIGTA326040
Supplier:  OriGene
Anti-ATF2 Rabbit Polyclonal Antibody
Catalog # ORIGTA326040
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Activating transcription factor 249
  • Antigen symbol:
    ATF249
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    ATF2
  • Modification:
    phospho-Ser472
  • Storage buffer:
    Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    The antiserum was produced against synthesized phosphopeptide derived from human ATF-2 around the phosphorylation site of serine 472 (A-L-SpQ-I).
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

  • Recommended Dilutions: Immunochemistry: 1:50-1:100 ELISA: 1:5000

Type: Primary
Antigen: ATF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat