Anti-VCP Mouse Monoclonal Antibody [clone: 4G9]
Catalog # ORIGTA328022
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Valosin containing protein
- Antigen symbol:VCP
- Clonality:Monoclonal
- Clone:4G9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:This antibody is provided in phosphate-buffered solution, pH 7,2, containing 0,09% sodium azide at 0,5 mg/ml.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Recombinant human valosin containing protein
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: VCP
Clonality: Monoclonal
Clone: 4G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human