Anti-Slc17a6 Mouse Monoclonal Antibody [clone: S29-29]
ORIGTA326590
: OriGene
- Antibody type:Primary
- Antigen symbol:Solute carrier family 17, member 6
- Clonality:Monoclonal
- Clone:S29-29
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:501 to 582
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2.
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Type: Primary
Antigen: Slc17a6
Clonality: Monoclonal
Clone: S29-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat