Order Entry
Export
ContactUsLinkComponent
Anti-Lrp4 Mouse Monoclonal Antibody [clone: S207-27]
Anti-Lrp4 Mouse Monoclonal Antibody [clone: S207-27]
Catalog # ORIGTA326569
Supplier:  OriGene
Anti-Lrp4 Mouse Monoclonal Antibody [clone: S207-27]
Catalog # ORIGTA326569
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    LRP4
  • Clonality:
    Monoclonal
  • Clone:
    S207-27
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS pH 7,4, 50% glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Fusion protein amino acids 26-
  • Purification:
    purified by Protein G chromatography
  • Size:
    100µg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1?g/ml

Type: Primary
Antigen: Lrp4
Clonality: Monoclonal
Clone: S207-27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2A
Reactivity: Mouse, Rat