Anti-Kcnc4 Mouse Monoclonal Antibody [clone: S72-16]
Catalog # ORIGTA326558
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:KCNC4
- Clonality:Monoclonal
- Clone:S72-16
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:175 to 192
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide amino acids 175-192 of rat Kv3,4,
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml
Type: Primary
Antigen: Kcnc4
Clonality: Monoclonal
Clone: S72-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat.