Anti-Grin2a Mouse Monoclonal Antibody [clone: S327A-38]
ORIGTA326538
: OriGene
- Antibody type:Primary
- Antigen symbol:GRIN2A
- Clonality:Monoclonal
- Clone:S327A-38
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:75 to 325
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 75-325 (extracellular N-terminus) of rat GluN2A/NR2A
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: Grin2a
Clonality: Monoclonal
Clone: S327A-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat