Order Entry
Export
ContactUsLinkComponent
Anti-Gabbr1 Mouse Monoclonal Antibody [clone: S93A-49]
Anti-Gabbr1 Mouse Monoclonal Antibody [clone: S93A-49]
  ORIGTA326530
 :  OriGene
Anti-Gabbr1 Mouse Monoclonal Antibody [clone: S93A-49]
  ORIGTA326530
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    GABBR1
  • Clonality:
    Monoclonal
  • Clone:
    S93A-49
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    873 to 977
  • Storage buffer:
    PBS pH 7,4, 50% glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Fusion protein amino acids 873-977 (cytoplasmic C-terminus) of rat GABA(B)R1
  • Purification:
    purified by Protein G chromatography
  • Size:
    100µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: Gabbr1
Clonality: Monoclonal
Clone: S93A-49
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat