Anti-Cask Mouse Monoclonal Antibody [clone: S56A-50]
Catalog # ORIGTA326522
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:Cask
- Clonality:Monoclonal
- Clone:S56A-50
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Zebrafish,Xenopus,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:318 to 415
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 318-415 of rat CASK/Lin2
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: Cask
Clonality: Monoclonal
Clone: S56A-50
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Xenopus, zebraFish