Anti-Abcc8 Mouse Monoclonal Antibody [clone: S289-16]
Catalog # ORIGTA326508
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family C, member 8
- Antigen symbol:ABCC8
- Clonality:Monoclonal
- Clone:S289-16
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Hamster,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:1548 to 1582
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 1548-1582 (cytoplasmic C-terminus) of rat SUR1
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml
Type: Primary
Antigen: Abcc8
Clonality: Monoclonal
Clone: S289-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Mouse, Rat