Order Entry
Export
ContactUsLinkComponent
Anti-Kcnu1 Mouse Monoclonal Antibody [clone: S2-16]
Anti-Kcnu1 Mouse Monoclonal Antibody [clone: S2-16]
Catalog # ORIGTA326490
Supplier:  OriGene
Anti-Kcnu1 Mouse Monoclonal Antibody [clone: S2-16]
Catalog # ORIGTA326490
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    KCNU1
  • Clonality:
    Monoclonal
  • Clone:
    S2-16
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    1052 to 1121
  • Storage buffer:
    PBS pH 7,4, 50% glycerol, 0,09% sodium azide
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Fusion protein amino acids 1052-1121 of mouse Slo3;
  • Purification:
    purified by Protein G chromatography
  • Size:
    100µg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml

Type: Primary
Antigen: Kcnu1
Clonality: Monoclonal
Clone: S2-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse