Anti-KCNA6 Mouse Monoclonal Antibody [clone: S19-36]
ORIGTA326483
: OriGene
- Antibody type:Primary
- Antigen symbol:KCNA6
- Clonality:Monoclonal
- Clone:S19-36
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG3
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:507 to 525
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Sequence:RERRSSYLPTPHRAYAEKR
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide 507-525 (RERRSSYLPTPHRAYAEKR, cytoplasmic C-terminus) of rat Kv1.6,
- Purification:purified by PEG
- Size:100µg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml
Type: Primary
Antigen: KCNA6
Clonality: Monoclonal
Clone: S19-36
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse, Rat