Anti-Asic1 Mouse Monoclonal Antibody [clone: S271-44]
ORIGTA326467
: OriGene
- Antibody type:Primary
- Antigen name:Acid sensing (proton gated) ion channel 1
- Antigen symbol:ASCI1
- Clonality:Monoclonal
- Clone:S271-44
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:ACCN2
- Amino acid number:460 to 526
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 460-526 (Cytoplasmic C-terminus) of mouse ASIC1,
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Type: Primary
Antigen: Asic1
Clonality: Monoclonal
Clone: S271-44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat