Anti-DLG4 Mouse Monoclonal Antibody [clone: S28-43]
Catalog # ORIGTA326433
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:DLG4
- Clonality:Monoclonal
- Clone:S28-43
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 77-200 of human PSD95/SAP90
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: DLG4
Clonality: Monoclonal
Clone: S28-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat