Anti-KCNMB3 Mouse Monoclonal Antibody [clone: S40B-18]
Catalog # ORIGTA326402
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:KCNMB3
- Clonality:Monoclonal
- Clone:S40B-18
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 1-49 (unique N-terminus) of mouse BKBeta3a
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml
Type: Primary
Antigen: KCNMB3
Clonality: Monoclonal
Clone: S40B-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat