Anti-CNTN1 Mouse Monoclonal Antibody [clone: S73-20]
Catalog # ORIGTA326400
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CNTN1
- Clonality:Monoclonal
- Clone:S73-20
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:21 to 37
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Sequence:D/EFTWHRRYGHGVSEEDKC
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide amino acids 21-37 (D/EFTWHRRYGHGVSEEDKC) extracellular domain of rat contactin,
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 0.1-1?g/ml; Immunofluorescence: 1-10?g/ml
Type: Primary
Antigen: CNTN1
Clonality: Monoclonal
Clone: S73-20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat