Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: AC-16]
Catalog # ORIGTA326371
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HSP90AB1
- Clonality:Monoclonal
- Clone:AC-16
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Plant,Rabbit,Fungus,Insect,Chicken,Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Heat shock protein 900 from the water mold Achyla ambisexualis
- Purification:purified by Protein G chromatography
- Pk:200 µG
Specifications
About this item
Recommended Dilutions: 1?g/ml
Type: Primary
Antigen: HSP90AB1
Clonality: Monoclonal
Clone: AC-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Achlya, Chicken, Human, Mouse, Rabbit, Rat, sf9 cell line, wheat germ