Order Entry
Export
ContactUsLinkComponent
Anti-SYP Rabbit Monoclonal Antibody [clone: MRQ-40]
Anti-SYP Rabbit Monoclonal Antibody [clone: MRQ-40]
Catalog # ORIGTA327740
Supplier:  OriGene
Anti-SYP Rabbit Monoclonal Antibody [clone: MRQ-40]
Catalog # ORIGTA327740
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Synaptophysin
  • Antigen symbol:
    SYP
  • Clonality:
    Monoclonal
  • Clone:
    MRQ-40
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    The antibody is diluted in tris buffered saline, pH 7,3-7.7, with 1% BSA and <1% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Purification:
    purified by affinity chromatography
  • Pk:
    1 mL

Specifications

About this item

Type: Primary
Antigen: SYP
Clonality: Monoclonal
Clone: MRQ-40
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human