Order Entry
Export
ContactUsLinkComponent
 
Anti-TBX1 Rabbit Polyclonal Antibody
Catalog # ORIGTA325103
Supplier:  OriGene
Anti-TBX1 Rabbit Polyclonal Antibody
Catalog # ORIGTA325103
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TBX1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    near the C-terminus
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    327 to 356
  • Storage buffer:
    Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
  • Molecular weight:
    43.13 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    This TBX1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 327-356 amino acids from the C-terminal region of human TBX1.
  • Purification:
    purified by Protein A chromatography, followed by affinity purification
  • Size:
    400 μl
  • Pk:
    400 µl

Specifications

About this item

  • Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:10-50; Immunochemistry: 1:50-100

Type: Primary
Antigen: TBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human