Order Entry
Export
ContactUsLinkComponent
Anti-ZAP70 Rabbit Polyclonal Antibody
Anti-ZAP70 Rabbit Polyclonal Antibody
Catalog # ORIGTA323317
Supplier:  OriGene
Anti-ZAP70 Rabbit Polyclonal Antibody
Catalog # ORIGTA323317
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    ZAP70
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    317 to 321
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Molecular weight:
    69 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Peptide sequence around aa.317~321 (S-P-Y-S-D) derived from Human ZAP70.
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100

Type: Primary
Antigen: ZAP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse