Anti-SFTPA1 Rabbit Polyclonal Antibody
ORIGTA323274
: OriGene
- Antibody type:Primary
- Antigen name:Surfactant Protein A1
- Antigen symbol:SFTPA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:223 to 236
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:28 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to a region derived from 223-236 amino acids of human surfactant protein A1
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:200-1000
Type: Primary
Antigen: SFTPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat