Anti-HK2 Rabbit Polyclonal Antibody
Catalog # ORIGTA325030
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Form:Liquid
- Amino acid number:453 to 483
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:102.38 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This HK2 (Hexokinase II) antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 453-483 amino acids from the Central region of human HK2 (Hexokinase II).
- Purification:purified by saturated ammonium sulfate precipitation followed by dialysis against PBS
- Size:400 μl
- Pk:400 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: HK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human