Anti-PFKL Rabbit Polyclonal Antibody
ORIGTA325027
: OriGene
- Antibody type:Primary
- Antigen symbol:PFKL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Liquid
- Amino acid number:669 to 699
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:85.02 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This PFKL antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 669-699 amino acids from the C-terminal region of human PFKL.
- Purification:purified by saturated ammonium sulfate precipitation followed by dialysis against PBS
- Size:400 μl
- Pk:400 µl
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50-100
Type: Primary
Antigen: PFKL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse