Anti-TIMP1 Rabbit Polyclonal Antibody
Catalog # ORIGTA322880
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:TIMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Amino acid number:corresponding to a region between amino acids 24 to 207
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein corresponding to a region derived from 24-207 amino acids of human TIMP metallopeptidase inhibitor 1
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: ELISA: 1:1000-2000; Immunochemistry: 1:15-50
Type: Primary
Antigen: TIMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat