Order Entry
Export
ContactUsLinkComponent
Anti-YWHAB Rabbit Polyclonal Antibody
Anti-YWHAB Rabbit Polyclonal Antibody
  ORIGTA322853
 :  OriGene
Anti-YWHAB Rabbit Polyclonal Antibody
  ORIGTA322853
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    YWHAB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Amino acid number:
    72 to 87
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Molecular weight:
    28 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Synthetic peptide corresponding to a region derived from 72-87 amino acids of Human tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta polypeptide
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Recommended Dilutions: ELISA: 1:1000-5000; Western Blot: 1:500-2000

Type: Primary
Antigen: YWHAB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat