Anti-TNFRSF11A Rat Polyclonal Antibody [clone: R12-31]
Catalog # ORIGTA320494
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:TNFRSF11A
- Clonality:Polyclonal
- Clone:R12-31
- Conjugation:Unconjugated
- Host:Rat
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TNFRSF11A
Clonality: Polyclonal
Clone: R12-31
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse