Anti-SPN Rat Polyclonal Antibody [clone: eBioR2/60]
Catalog # ORIGTA320466
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:SPN
- Clonality:Polyclonal
- Clone:eBioR2/60
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SPN
Clonality: Polyclonal
Clone: eBioR2/60
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse