Anti-LAG3 Mouse Polyclonal Antibody [clone: CNA.42]
Catalog # ORIGTA320408
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:LAG3
- Clonality:Polyclonal
- Clone:CNA.42
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:25 μg
- Pk:25 µG
Specifications
About this item
Type: Primary
Antigen: LAG3
Clonality: Polyclonal
Clone: CNA.42
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human