Anti-CDX2 Rabbit Polyclonal Antibody
Catalog # ORIGTA323197
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Caudal type homeobox 2
- Antigen symbol:CDX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:12 to 16
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:34 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Peptide sequence around aa.12~16(S-M-Y-P-S) derived from Human Cdx2
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1000
Type: Primary
Antigen: CDX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat