Order Entry
Export
ContactUsLinkComponent
 
Anti-POLR2I Rabbit Polyclonal Antibody
Catalog # ORIGTA322526
Supplier:  OriGene
Anti-POLR2I Rabbit Polyclonal Antibody
Catalog # ORIGTA322526
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    POLR2I
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS pH 7,3, 0,05% NaN3, 50% glycerol
  • Molecular weight:
    15 kDa
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Full length fusion protein
  • Purification:
    purified by antigen affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:500-2000

Type: Primary
Antigen: POLR2I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat