Anti-NCOR1 Rabbit Polyclonal Antibody
ORIGTA322513
: OriGene
- Antibody type:Primary
- Antigen symbol:NCOR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Amino acid number:840 to 894
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to a region derived from 840-894 amino acids of Human nuclear receptor corepressor 1
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: ELISA: 1:2000-10000; Immunochemistry: 1:100-300
Type: Primary
Antigen: NCOR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat