Anti-HSPA9 Rabbit Polyclonal Antibody
ORIGTA321626
: OriGene
- Antibody type:Primary
- Antigen symbol:HSPA9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:74 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein corresponding to C terminal 300 amino acids of human heat shock 70kDa protein 9 (mortalin)
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-2000
Type: Primary
Antigen: HSPA9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat