Anti-SOCS3 Rabbit Polyclonal Antibody
Catalog # ORIGTA321613
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:SOCS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length fusion protein
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: ELISA: 1:1000-5000; Immunochemistry: 1:25-100
Type: Primary
Antigen: SOCS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat