Anti-pT-PN11 Rabbit Polyclonal Antibody
Catalog # ORIGTA321539
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:pT-PN11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Modification:phospho-Tyr542
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:68 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:PepT-ide sequence around phosphorylation site of tyrosine 542 (H-E-Y(p)-T-N) derived from Human SHp2.
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50-100
Type: Primary
Antigen: pT-PN11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat