Anti-TACC2 Mouse Monoclonal Antibody [clone: 5H1]
ORIGCF804280
: OriGene
- Antibody type:Primary
- Antigen symbol:TACC2
- Clonality:Monoclonal
- Clone:5H1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:727 to 1026
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 727-1026 of human TACC2 (NP_008928) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: TACC2
Clonality: Monoclonal
Clone: 5H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human