Order Entry
Export
ContactUsLinkComponent
 
Anti-BrdU Rat Monoclonal Antibody [clone: 3B2]
  ORIGTA190091
 :  OriGene
Anti-BrdU Rat Monoclonal Antibody [clone: 3B2]
  ORIGTA190091
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen symbol:
    BRDU
  • Clonality:
    Monoclonal
  • Clone:
    3B2
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Bromodeoxyuridine coupled to keyhole limpet hemocyanin (KLH).
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: 3B2
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human