Anti-KRT19 Mouse Monoclonal Antibody [clone: 6E8]
ORIGCF803998
: OriGene
- Antibody type:Primary
- Antigen symbol:KRT19
- Clonality:Monoclonal
- Clone:6E8
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:240 to 390
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:43.9 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 240-390 of human KRT19 (NP_002267) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Recommended Dilutions: Immunochemistry: 1:150; Western Blot: 1:200-1:1000
Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: 6E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human