Anti-IL23R Mouse Monoclonal Antibody [clone: 5H9]
Catalog # ORIGCF807494
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:IL23R
- Clonality:Monoclonal
- Clone:5H9
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:57 to 355
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:69 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 57-355 of human IL23R(NP_653302) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: IL23R
Clonality: Monoclonal
Clone: 5H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human