Anti-DVL2 Mouse Monoclonal Antibody [clone: 7F10]
Catalog # ORIGCF806918
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Dishevelled segment polarity protein 2
- Antigen symbol:DVL2
- Clonality:Monoclonal
- Clone:7F10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:1 to 320
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:78.8 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 1-320 of human DVL2(NP_004413) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: DVL2
Clonality: Monoclonal
Clone: 7F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human