Anti-RSPO3 Mouse Monoclonal Antibody [clone: 3D5]
ORIGCF804633
: OriGene
- Antibody type:Primary
- Antigen symbol:RSPO3
- Clonality:Monoclonal
- Clone:3D5
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:22 to 272
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:30.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 22-272 of human RSPO3 (NP_116173) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Recommended Dilutions: Immunochemistry: 1:150
Type: Primary
Antigen: RSPO3
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human