Anti-APC Mouse Monoclonal Antibody [clone: 1B12]
Catalog # ORIGCF803682
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:adenomatous polyposis coli
- Antigen symbol:APC
- Clonality:Monoclonal
- Clone:1B12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:955 to 1382
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:311.5 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 955-1382 of human APC (NP_001120983) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Immunochemistry: 1:150
Type: Primary
Antigen: APC
Clonality: Monoclonal
Clone: 1B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human