Anti-IDO2 Mouse Monoclonal Antibody [clone: 8B5]
ORIGCF806618
: OriGene
- Antibody type:Primary
- Antigen name:Indoleamine 2,3-dioxygenase 2
- Antigen symbol:IDO2
- Clonality:Monoclonal
- Clone:8B5
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human IDO2 (NP_919270) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: IDO2
Clonality: Monoclonal
Clone: 8B5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human