Anti-NARS2 Mouse Monoclonal Antibody [clone: 10C10]
Catalog # ORIGCF806497
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:NARS2
- Clonality:Monoclonal
- Clone:10C10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:53.9 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human NARS2 (NP_078954) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: NARS2
Clonality: Monoclonal
Clone: 10C10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human