Anti-CD80 Armenian Hamster Polyclonal Antibody [clone: 16-10A1]
Catalog # ORIGTA320291
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:CD80
- Clonality:Polyclonal
- Clone:16-10A1
- Conjugation:Unconjugated
- Host:Armenian hamster
- Reactivity:Pig,Dog,Mouse
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:50 μg
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CD80
Clonality: Polyclonal
Clone: 16-10A1
Conjugation: Unconjugated
Epitope:
Host: Armenian Hamster
Isotype:
Reactivity: canine, Mouse, Pig