Anti-IL-10 Rabbit Polyclonal Antibody
Catalog # ORIGTA319291
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:IL-10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:None
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:The whole rabbit serum was prepared by repeated immunizations with human IL-10.
- Pk:1 ml
Specifications
About this item
Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000
Type: Primary
Antigen: IL-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human