Order Entry
Export
ContactUsLinkComponent
Anti-BIN1 Mouse Monoclonal Antibody [clone: 12A]
Anti-BIN1 Mouse Monoclonal Antibody [clone: 12A]
Catalog # ORIGTA319579
Supplier:  OriGene
Anti-BIN1 Mouse Monoclonal Antibody [clone: 12A]
Catalog # ORIGTA319579
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    BIN1
  • Clonality:
    Monoclonal
  • Clone:
    12A
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Anti-BIN1 (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with 12A exon BIN1 protein followed by hybridoma development.
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: ELISA: 1:5000-1:50000; Western Blot: 1:500-1:1500

Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 12A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human